Product Details
- SNP ID
-
rs150797486
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:52940341 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGTGTTGGCCACTGCGGAGTCGC[A/G]CCGGGTGGCCGGCCGCACCTACAGC
- Phenotype
-
MIM: 600730
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NPBWR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs33977775] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NPBWR1
- Gene Name
- neuropeptides B/W receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005285.3 |
434 |
Missense Mutation |
CAC,CGC |
H145R |
NP_005276.2 |
View Full Product Details