Product Details

SNP ID
rs150997442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123249176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTGAACATTTCTTCTACGAAG[A/G]AAGCGCATTTTGAAAAATGGTCCTA
Phenotype
MIM: 604764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf76 PubMed Links

Gene Details

Gene
C8orf76
Gene Name
chromosome 8 open reading frame 76
There are no transcripts associated with this gene.

Gene
ZHX1
Gene Name
zinc fingers and homeoboxes 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017926.2 4475 UTR 3 NP_001017926.1
NM_007222.4 4475 UTR 3 NP_009153.3
Gene
ZHX1-C8orf76
Gene Name
ZHX1-C8orf76 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204180.1 4475 Intron NP_001191109.1

View Full Product Details