Product Details

SNP ID
rs137862169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132167039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGACCCCCCTGAGAGGTTCTGC[G/T]CCCATGTAAGTCCACTTACTGCTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NTNG2 PubMed Links

Gene Details

Gene
NTNG2
Gene Name
netrin G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032536.2 1005 Missense Mutation GCC,TCC A70S NP_115925.2
XM_006717304.3 1005 Missense Mutation GCC,TCC A70S XP_006717367.1
XM_011519094.2 1005 Missense Mutation GCC,TCC A70S XP_011517396.1
XM_011519096.2 1005 Missense Mutation GCC,TCC A70S XP_011517398.1
XM_011519097.2 1005 Missense Mutation GCC,TCC A70S XP_011517399.1
XM_011519098.2 1005 Missense Mutation GCC,TCC A70S XP_011517400.1
XM_011519099.2 1005 Missense Mutation GCC,TCC A70S XP_011517401.1
XM_011519100.2 1005 Missense Mutation GCC,TCC A70S XP_011517402.1
XM_011519102.2 1005 Missense Mutation GCC,TCC A70S XP_011517404.1
XM_011519103.2 1005 Missense Mutation GCC,TCC A70S XP_011517405.1
XM_011519104.2 1005 Missense Mutation GCC,TCC A70S XP_011517406.1
XM_011519105.2 1005 Missense Mutation GCC,TCC A70S XP_011517407.1
XM_011519106.2 1005 Missense Mutation GCC,TCC A70S XP_011517408.1
XM_011519107.2 1005 Missense Mutation GCC,TCC A70S XP_011517409.1
XM_011519108.2 1005 Missense Mutation GCC,TCC A70S XP_011517410.1
XM_011519109.2 1005 Missense Mutation GCC,TCC A70S XP_011517411.1
XM_011519110.2 1005 Missense Mutation GCC,TCC A70S XP_011517412.1
XM_011519112.2 1005 Intron XP_011517414.1
XM_011519113.2 1005 Intron XP_011517415.1
XM_017015212.1 1005 Missense Mutation GCC,TCC A70S XP_016870701.1
XM_017015213.1 1005 Missense Mutation GCC,TCC A70S XP_016870702.1
XM_017015214.1 1005 Missense Mutation GCC,TCC A70S XP_016870703.1
XM_017015215.1 1005 Intron XP_016870704.1
XM_017015216.1 1005 Missense Mutation GCC,TCC A70S XP_016870705.1

View Full Product Details