Product Details
- SNP ID
-
rs137921767
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21350503 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTCATCAGGGGAGTCCCTCCCA[C/T]CCACACCTCCTGCATCACACAGGCT
- Phenotype
-
MIM: 147566
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA6
PubMed Links
Gene Details
- Gene
- IFNA6
- Gene Name
- interferon alpha 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021002.2 |
385 |
Missense Mutation |
ATG,GTG |
M129V |
NP_066282.1 |
View Full Product Details