Product Details

SNP ID
rs138422440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33255296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTCTGCTCCACTGTGTCACACT[C/T]GGCTAGGAATGCCTAGAAAATACAC
Phenotype
MIM: 601497 MIM: 610900 MIM: 613929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172415.1 1048 Missense Mutation AAG,GAG K206E NP_001165886.1
NM_004323.5 1048 Missense Mutation AAG,GAG K321E NP_004314.5
Gene
CHMP5
Gene Name
charged multivesicular body protein 5
There are no transcripts associated with this gene.

Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details