Product Details

SNP ID
rs138626161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91722888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTAAGCTCTGTACATGATTCT[C/T]AACAAAAATAACAATACCGTGTTCT
Phenotype
MIM: 602337
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ROR2 PubMed Links

Gene Details

Gene
ROR2
Gene Name
receptor tyrosine kinase like orphan receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318204.1 2511 Intron NP_001305133.1
NM_004560.3 2511 UTR 3 NP_004551.2
XM_005252008.4 2511 UTR 3 XP_005252065.1
XM_005252009.3 2511 UTR 3 XP_005252066.1
XM_006717121.3 2511 UTR 3 XP_006717184.1
XM_017014762.1 2511 UTR 3 XP_016870251.1
XM_017014763.1 2511 UTR 3 XP_016870252.1

View Full Product Details