Product Details

SNP ID
rs139024531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36578291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTAATCAATTGTCCCAGCTCCA[G/T]AGCTGCATTTCCATCACCTGTTACC
Phenotype
MIM: 607025
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MELK PubMed Links

Gene Details

Gene
MELK
Gene Name
maternal embryonic leucine zipper kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256685.1 Intron NP_001243614.1
NM_001256687.1 Intron NP_001243616.1
NM_001256688.1 Intron NP_001243617.1
NM_001256689.1 Intron NP_001243618.1
NM_001256690.1 Intron NP_001243619.1
NM_001256691.1 Intron NP_001243620.1
NM_001256692.1 Intron NP_001243621.1
NM_001256693.1 Intron NP_001243622.1
NM_014791.3 Intron NP_055606.1
XM_011518076.2 Intron XP_011516378.1
XM_011518077.1 Intron XP_011516379.1
XM_011518078.2 Intron XP_011516380.1
XM_011518079.1 Intron XP_011516381.1
XM_011518080.1 Intron XP_011516382.1
XM_011518081.2 Intron XP_011516383.1
XM_011518082.2 Intron XP_011516384.1
XM_011518083.2 Intron XP_011516385.1
XM_011518084.2 Intron XP_011516386.1
XM_011518085.1 Intron XP_011516387.1
XM_011518086.2 Intron XP_011516388.1

View Full Product Details