Product Details

SNP ID
rs139074626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38395800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTAGCCTCCAGGGCAGGACCGCC[C/T]GCTACTCCTCGGCAGCAGCCCTCCC
Phenotype
MIM: 100670
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH1B1 PubMed Links

Gene Details

Gene
ALDH1B1
Gene Name
aldehyde dehydrogenase 1 family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000692.4 205 Missense Mutation CGC,TGC R18C NP_000683.3
XM_011517802.2 205 Missense Mutation CGC,TGC R18C XP_011516104.1

View Full Product Details