Product Details

SNP ID
rs139206714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127786779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGCTGATGGAAAACGAGAAGGA[A/G]GGGGTGAGTACGGATCGGGCGTGCG
Phenotype
MIM: 603251 MIM: 613405 MIM: 604722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK9 PubMed Links

Gene Details

Gene
CDK9
Gene Name
cyclin dependent kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261.3 294 Silent Mutation GAA,GAG E57E NP_001252.1
XM_017014184.1 294 UTR 5 XP_016869673.1
Gene
MIR2861
Gene Name
microRNA 2861
There are no transcripts associated with this gene.

Gene
MIR3960
Gene Name
microRNA 3960
There are no transcripts associated with this gene.

Gene
SH2D3C
Gene Name
SH2 domain containing 3C
There are no transcripts associated with this gene.

View Full Product Details