Product Details

SNP ID
rs139565812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86946144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGAGCAGCGCCGCCTTGGGCAT[A/T]GCCAGCATGTCCTCAATGACGGTGC
Phenotype
MIM: 139185
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GAS1 PubMed Links

Gene Details

Gene
GAS1
Gene Name
growth arrest specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002048.2 1046 Silent Mutation GCA,GCT A212A NP_002039.2
Gene
GAS1RR
Gene Name
GAS1 adjacent regulatory RNA
There are no transcripts associated with this gene.

View Full Product Details