Product Details

SNP ID
rs140444141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135663387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCAGGAGTTCGATCCCCACAC[C/T]GTTATGCAGAGGAACTACAACGTGG
Phenotype
MIM: 612903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCN9 PubMed Links

Gene Details

Gene
LCN9
Gene Name
lipocalin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001676.1 66 Silent Mutation ACC,ACT T22T NP_001001676.1
XM_017014712.1 66 Silent Mutation ACC,ACT T22T XP_016870201.1

View Full Product Details