Product Details
- SNP ID
-
rs140515102
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:27326924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTACTATCTAAGAGTTTTCCCAA[A/G]AAAACCATCTGACAATTAGGGAGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MOB3B
PubMed Links
Gene Details
- Gene
- MOB3B
- Gene Name
- MOB kinase activator 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024761.4 |
4810 |
UTR 3 |
|
|
NP_079037.3 |
View Full Product Details