Product Details
- SNP ID
-
rs140752754
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112997700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGTACTGTGGCAAAAAACTTTC[C/T]GACATTGCTTACACTGGTAGGATTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF883
PubMed Links
Gene Details
- Gene
- ZNF883
- Gene Name
- zinc finger protein 883
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001101338.1 |
1799 |
Missense Mutation |
CAG,CGG |
Q187R |
NP_001094808.1 |
View Full Product Details