Product Details

SNP ID
rs140766699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128705371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGCCGGGCCATCCAGAAAGAGCT[A/G]AAGATCAAGGAGGGGGTGGAGAACC
Phenotype
MIM: 610714 MIM: 600960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 576 Silent Mutation CTA,CTG L31L NP_001304855.1
NM_013355.4 576 Silent Mutation CTA,CTG L31L NP_037487.2
XM_005251946.3 576 Silent Mutation CTA,CTG L28L XP_005252003.1
XM_006717080.2 576 Silent Mutation CTA,CTG L38L XP_006717143.1
XM_017014649.1 576 Silent Mutation CTA,CTG L38L XP_016870138.1
XM_017014650.1 576 UTR 5 XP_016870139.1
Gene
SET
Gene Name
SET nuclear proto-oncogene
There are no transcripts associated with this gene.

View Full Product Details