Product Details

SNP ID
rs140797044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34254498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGAACCATGATTTCATCCAGC[C/T]GGGTCACAAAATCTTCAAAATCCTG
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 2713 Missense Mutation CAG,CGG Q1330R NP_919289.2
XM_005251451.2 2713 Missense Mutation CAG,CGG Q969R XP_005251508.1
XM_006716764.2 2713 Missense Mutation CAG,CGG Q872R XP_006716827.1
XM_011517861.1 2713 Missense Mutation CAG,CGG Q1361R XP_011516163.1
XM_011517863.2 2713 Missense Mutation CAG,CGG Q1330R XP_011516165.1
XM_017014697.1 2713 Missense Mutation CAG,CGG Q1330R XP_016870186.1
XM_017014698.1 2713 Missense Mutation CAG,CGG Q1041R XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details