Product Details
- SNP ID
-
rs141303226
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:33240218 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATCCCAGGATCAGCATGGCCGTC[C/T]GCCAGTGGGTAATCGCCCTGGCCTT
- Phenotype
-
MIM: 613929
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPINK4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs706107] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPINK4
- Gene Name
- serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.
View Full Product Details