Product Details

SNP ID
rs141381082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136853153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAACCACTGCAGGAGCCCTGGCC[A/T]GGCCGTGTGCAACTTCGTGTGTGAC
Phenotype
MIM: 605107 MIM: 610167
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf172 PubMed Links

Gene Details

Gene
C9orf172
Gene Name
chromosome 9 open reading frame 172
There are no transcripts associated with this gene.

Gene
EDF1
Gene Name
endothelial differentiation related factor 1
There are no transcripts associated with this gene.

Gene
MAMDC4
Gene Name
MAM domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206920.2 148 Missense Mutation CAG,CTG Q33L NP_996803.2
Gene
PHPT1
Gene Name
phosphohistidine phosphatase 1
There are no transcripts associated with this gene.

View Full Product Details