Product Details

SNP ID
rs141434421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122800350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGCTGACCTCTGCTTCGCCTC[C/T]GTCACTGTGCCCAAGATGTTGGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1K1 PubMed Links

Gene Details

Gene
OR1K1
Gene Name
olfactory receptor family 1 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080859.1 228 Silent Mutation TCC,TCT S76S NP_543135.1

View Full Product Details