Product Details

SNP ID
rs141658118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128345443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCTGGGCATGGCCAAGCTCGG[A/T]GTGGAGGCAGCCCTCATCAACACCA
Phenotype
MIM: 604194
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC27A4 PubMed Links

Gene Details

Gene
SLC27A4
Gene Name
solute carrier family 27 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005094.3 608 Silent Mutation GGA,GGT G150G NP_005085.2
XM_017014222.1 608 Silent Mutation GGA,GGT G150G XP_016869711.1

View Full Product Details