Product Details

SNP ID
rs142216050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98290344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTTGCAGATGTTAAGGAAGAC[A/G]TCCCATTTCCGTTCCTCTTCTTTGT
Phenotype
MIM: 607340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABBR2 PubMed Links

Gene Details

Gene
GABBR2
Gene Name
gamma-aminobutyric acid type B receptor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005458.7 2944 UTR 3 NP_005449.5
XM_005252316.4 2944 UTR 3 XP_005252373.1
XM_017015331.1 2944 UTR 3 XP_016870820.1
XM_017015332.1 2944 UTR 3 XP_016870821.1

View Full Product Details