Product Details

SNP ID
rs142366975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128343133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCCCTGTGTCCGCAGGCCACA[A/G]TGCTGCTTGGAGCCTCTCTGGTGGG
Phenotype
MIM: 612898 MIM: 604194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ4 PubMed Links

Gene Details

Gene
COQ4
Gene Name
coenzyme Q4
There are no transcripts associated with this gene.

Gene
SLC27A4
Gene Name
solute carrier family 27 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005094.3 159 Missense Mutation ATG,GTG M1V NP_005085.2
XM_017014222.1 159 Missense Mutation ATG,GTG M1V XP_016869711.1

View Full Product Details