Product Details
- SNP ID
-
rs142377644
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:100113049 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAGTCTCATTATGGCATATTCTCT[A/G]TAAGTATCACTGACTCCAGAAGGAC
- Phenotype
-
MIM: 243305
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
INVS
PubMed Links
Gene Details
- Gene
- INVS
- Gene Name
- inversin
View Full Product Details