Product Details

SNP ID
rs142633392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114331576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTCTCTTCCAGAGGGAGGCC[A/G]AGAACATGTTGCTCACCTGCTGTTC
Phenotype
MIM: 605729 MIM: 138600 MIM: 138610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKNA PubMed Links

Gene Details

Gene
AKNA
Gene Name
AT-hook transcription factor
There are no transcripts associated with this gene.

Gene
ORM1
Gene Name
orosomucoid 1
There are no transcripts associated with this gene.

Gene
ORM2
Gene Name
orosomucoid 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000608.2 454 Missense Mutation CAA,CGA Q113R NP_000599.1

View Full Product Details