Product Details

SNP ID
rs142806307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137301836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGTCATGTTCTGCAGCAGCGA[C/T]TGCAGCTCCTGCGTGTTGCCCTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXD3 PubMed Links

Gene Details

Gene
EXD3
Gene Name
exonuclease 3'-5' domain containing 3
There are no transcripts associated with this gene.

Gene
NRARP
Gene Name
NOTCH-regulated ankyrin repeat protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004354.2 416 Silent Mutation CAA,CAG Q31Q NP_001004354.1

View Full Product Details