Product Details

SNP ID
rs142846298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127455011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATCCTTAGGCAAAAGAAGCTG[G/T]GGCAGATGACATTCTCGACATCTCT
Phenotype
MIM: 610933 MIM: 180475 MIM: 194552
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRSAM1 PubMed Links

Gene Details

Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005373.3 1457 Missense Mutation GGG,GTG G29V NP_001005373.1
NM_001005374.3 1457 Missense Mutation GGG,GTG G29V NP_001005374.1
NM_001190723.2 1457 Missense Mutation GGG,GTG G29V NP_001177652.1
NM_138361.5 1457 Missense Mutation GGG,GTG G29V NP_612370.3
XM_006717316.3 1457 Missense Mutation GGG,GTG G29V XP_006717379.1
XM_017015283.1 1457 Missense Mutation GGG,GTG G29V XP_016870772.1
XM_017015284.1 1457 UTR 5 XP_016870773.1
Gene
RPL12
Gene Name
ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SNORA65
Gene Name
small nucleolar RNA, H/ACA box 65
There are no transcripts associated with this gene.

Gene
ZNF79
Gene Name
zinc finger protein 79
There are no transcripts associated with this gene.

View Full Product Details