Product Details

SNP ID
rs143617269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77177728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGTTTTCGAGTCGGTGGTCGT[A/G]GACGTGTTGAACCGGTTCTTGGGGG
Phenotype
MIM: 605978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS13A PubMed Links

Gene Details

Gene
VPS13A
Gene Name
vacuolar protein sorting 13 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018037.1 284 Silent Mutation GTA,GTG V8V NP_001018047.1
NM_001018038.2 284 Silent Mutation GTA,GTG V8V NP_001018048.1
NM_015186.3 284 Silent Mutation GTA,GTG V8V NP_056001.1
NM_033305.2 284 Silent Mutation GTA,GTG V8V NP_150648.2
Gene
VPS13A-AS1
Gene Name
VPS13A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details