Product Details

SNP ID
rs143635841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111363922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACAACAAATGGCAATATTTAAA[C/T]GATCCATTTTAGAAGTGGGAGATGA
Phenotype
MIM: 616694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0368 PubMed Links

Gene Details

Gene
KIAA0368
Gene Name
KIAA0368
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080398.1 Intron NP_001073867.1
XM_005251850.3 Intron XP_005251907.1
XM_005251851.4 Intron XP_005251908.1
XM_005251852.4 Intron XP_005251909.1
XM_005251853.3 Intron XP_005251910.1
XM_005251854.3 Intron XP_005251911.1
XM_011518422.2 Intron XP_011516724.1
XM_017014557.1 Intron XP_016870046.1

View Full Product Details