Product Details

SNP ID
rs143663154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33824141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCATAGAATATGCTTTTATCCT[A/G]TTTAGTTGATTACAAGATGCATATT
Phenotype
MIM: 612506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2R2 PubMed Links

Gene Details

Gene
UBE2R2
Gene Name
ubiquitin conjugating enzyme E2 R2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017811.3 Intron NP_060281.2
XM_005251496.2 Intron XP_005251553.1
XM_011517949.1 Intron XP_011516251.1
XM_017014860.1 Intron XP_016870349.1

View Full Product Details