Product Details

SNP ID
rs143979524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101422253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAACATGTTTATTTTAAAAAC[A/G]TTTCCATTTTCTACCTATATTGGTC
Phenotype
MIM: 612724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDOB PubMed Links

Gene Details

Gene
ALDOB
Gene Name
aldolase, fructose-bisphosphate B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000035.3 Intron NP_000026.2

View Full Product Details