Product Details

SNP ID
rs144093198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:2021516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATATTTCAGTTTTTACCATCCCA[A/C]TGGGAGATGCTTTATCACACATGGA
Phenotype
MIM: 600014
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMARCA2 PubMed Links

Gene Details

Gene
SMARCA2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289396.1 Intron NP_001276325.1
NM_001289397.1 Intron NP_001276326.1
NM_001289398.1 Intron NP_001276327.1
NM_001289399.1 Intron NP_001276328.1
NM_001289400.1 Intron NP_001276329.1
NM_003070.4 Intron NP_003061.3
NM_139045.3 Intron NP_620614.2

View Full Product Details