Product Details

SNP ID
rs144779571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93450500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTTTTTCACCGGGAGTATGGG[A/G]GCTTCTTTGGCCAAACCGTGCGCGT
Phenotype
MIM: 612265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM120A PubMed Links

Gene Details

Gene
FAM120A
Gene Name
family with sequence similarity 120A
There are no transcripts associated with this gene.

Gene
FAM120AOS
Gene Name
family with sequence similarity 120A opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322224.1 1546 Silent Mutation GCC,GCT A39A NP_001309153.1
NM_198841.2 1546 Silent Mutation GCC,GCT A221A NP_942138.2

View Full Product Details