Product Details
- SNP ID
-
rs145209418
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:135562401 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATCACCTCACTGTTGCCCACCCC[C/T]GAGGACAACCTGGAGATCGTTCTGC
- Phenotype
-
MIM: 173310
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PAEP
PubMed Links
Gene Details
- Gene
- PAEP
- Gene Name
- progestagen associated endometrial protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001018048.1 |
184 |
Silent Mutation |
CCC,CCT |
P46P |
NP_001018058.1 |
NM_001018049.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
NP_001018059.1 |
NM_002571.3 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
NP_002562.2 |
XM_011518745.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_011517047.1 |
XM_011518746.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_011517048.1 |
XM_011518747.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_011517049.1 |
XM_011518748.1 |
184 |
Silent Mutation |
CCC,CCT |
P46P |
XP_011517050.1 |
XM_011518749.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_011517051.1 |
XM_011518751.1 |
184 |
Silent Mutation |
CCC,CCT |
P46P |
XP_011517053.1 |
XM_011518752.2 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_011517054.1 |
XM_017014782.1 |
184 |
Intron |
|
|
XP_016870271.1 |
XM_017014783.1 |
184 |
Silent Mutation |
CCC,CCT |
P68P |
XP_016870272.1 |
View Full Product Details