Product Details
- SNP ID
-
rs145473718
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:83663404 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAGCCATATTTACGAGATATATA[C/T]ATATGTAACACAAGCTACCGCAGGA
- Phenotype
-
MIM: 605046
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBQLN1
PubMed Links
Gene Details
- Gene
- UBQLN1
- Gene Name
- ubiquilin 1
View Full Product Details