Product Details

SNP ID
rs145584417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122789391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCCTTGCTGGGAGCATCAGG[C/G]CTGGGTGGGGCAGTGAGTGCCTTTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5C1 PubMed Links

Gene Details

Gene
OR5C1
Gene Name
olfactory receptor family 5 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001923.1 459 Silent Mutation GGC,GGG G153G NP_001001923.1

View Full Product Details