Product Details

SNP ID
rs146126836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136850047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGACAAAGTGTCGGGCGACATGCA[A/G]AAGCAAGGCTGCGACTGTGAGTGTC
Phenotype
MIM: 610167 MIM: 610615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf172 PubMed Links

Gene Details

Gene
C9orf172
Gene Name
chromosome 9 open reading frame 172
There are no transcripts associated with this gene.

Gene
MAMDC4
Gene Name
MAM domain containing 4
There are no transcripts associated with this gene.

Gene
PHPT1
Gene Name
phosphohistidine phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135861.2 810 Silent Mutation CAA,CAG Q65Q NP_001129333.1
NM_001287342.1 810 Silent Mutation CAA,CAG Q67Q NP_001274271.1
NM_001287343.1 810 Silent Mutation CAA,CAG Q65Q NP_001274272.1
NM_014172.5 810 Silent Mutation CAA,CAG Q65Q NP_054891.2
Gene
RABL6
Gene Name
RAB, member RAS oncogene family-like 6
There are no transcripts associated with this gene.

View Full Product Details