Product Details
- SNP ID
-
rs146197742
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21166060 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGGTTTCATTCCTTCCTCCTTA[A/G]TCTTTCTTGAAAAATTTTTGATAAA
- Phenotype
-
MIM: 147584
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3750478] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNA21
- Gene Name
- interferon alpha 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002175.2 |
601 |
Silent Mutation |
CTA,TTA |
L185L |
NP_002166.2 |
View Full Product Details