Product Details
- SNP ID
-
rs146656324
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:101390738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCTTTAACGTAAGATTCCAAAC[A/G]ACTCTGGAGATCTTCAGGTGGTGTG
- Phenotype
-
MIM: 602938
MIM: 611854
MIM: 603132
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BAAT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10989486] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAAT
- Gene Name
- bile acid-CoA:amino acid N-acyltransferase
There are no transcripts associated with this gene.
- Gene
- MRPL50
- Gene Name
- mitochondrial ribosomal protein L50
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019051.2 |
|
Intron |
|
|
NP_061924.1 |
- Gene
- ZNF189
- Gene Name
- zinc finger protein 189
There are no transcripts associated with this gene.
View Full Product Details