Product Details

SNP ID
rs146712783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135498635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCCGTAAGCCTGGTTACTGG[C/T]CCTGTACACGGAGACAGCCTTCTGG
Phenotype
MIM: 614502 MIM: 611971 MIM: 614056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf116 PubMed Links

Gene Details

Gene
C9orf116
Gene Name
chromosome 9 open reading frame 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048265.1 236 Missense Mutation ACC,GCC T58A NP_001041730.1
NM_144654.2 236 Missense Mutation ACC,GCC T58A NP_653255.1
Gene
LOC101928525
Gene Name
uncharacterized LOC101928525
There are no transcripts associated with this gene.

Gene
MRPS2
Gene Name
mitochondrial ribosomal protein S2
There are no transcripts associated with this gene.

Gene
PPP1R26
Gene Name
protein phosphatase 1 regulatory subunit 26
There are no transcripts associated with this gene.

View Full Product Details