Product Details

SNP ID
rs147170359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4797690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTACTTGACAGTAGTAATTAAA[C/T]CTTTTCTTTTCCCTGTATCCCTAAT
Phenotype
MIM: 611405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RCL1 PubMed Links

Gene Details

Gene
RCL1
Gene Name
RNA terminal phosphate cyclase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286699.1 Intron NP_001273628.1
NM_001286700.1 Intron NP_001273629.1
NM_001286701.1 Intron NP_001273630.1
NM_005772.4 Intron NP_005763.3
XM_006716715.3 Intron XP_006716778.1
XM_011517673.2 Intron XP_011515975.1
XM_017014176.1 Intron XP_016869665.1

View Full Product Details