Product Details

SNP ID
rs147275093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137612981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTAGTGGAGGAGGGTTACTTCAA[C/G]AGTTCCCTGTCGCTGGCAGACAAGG
Phenotype
MIM: 607001
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARRDC1 PubMed Links

Gene Details

Gene
ARRDC1
Gene Name
arrestin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317968.1 281 Silent Mutation AAC,AAG N68K NP_001304897.1
NM_152285.3 281 Silent Mutation AAC,AAG N68K NP_689498.1
XM_005266119.1 281 Missense Mutation AAC,AAG N68K XP_005266176.1
XM_006717320.3 281 Missense Mutation AAC,AAG N75K XP_006717383.1
XM_006717321.3 281 Missense Mutation AAC,AAG N75K XP_006717384.1
XM_006717322.3 281 Missense Mutation AAC,AAG N75K XP_006717385.1
XM_017015288.1 281 Missense Mutation AAC,AAG N75K XP_016870777.1
Gene
ARRDC1-AS1
Gene Name
ARRDC1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
EHMT1
Gene Name
euchromatic histone lysine methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details