Product Details

SNP ID
rs147346447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37438503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCAGGGCTCCAGCATGTATCA[A/G]AGATAAGCCCCAAAAGCAAAGTCCA
Phenotype
MIM: 604296 MIM: 616590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRHPR PubMed Links

Gene Details

Gene
GRHPR
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012203.1 4249 Intron NP_036335.1
XM_005251631.1 4249 Intron XP_005251688.1
XM_011518073.2 4249 Intron XP_011516375.1
XM_017015320.1 4249 Intron XP_016870809.1
XM_017015321.1 4249 Intron XP_016870810.1
XM_017015322.1 4249 Intron XP_016870811.1
XM_017015323.1 4249 Intron XP_016870812.1
Gene
ZBTB5
Gene Name
zinc finger and BTB domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014872.2 4249 UTR 3 NP_055687.1
XM_005251634.1 4249 UTR 3 XP_005251691.1

View Full Product Details