Product Details
- SNP ID
-
rs147454118
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:114061484 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGACGAAGTTGTTACCGTTGCCCA[C/T]GCAGCCGCCGTACTGGAAAGTCTCA
- Phenotype
-
MIM: 176870
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMBP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112109765] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMBP
- Gene Name
- alpha-1-microglobulin/bikunin precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001633.3 |
1056 |
Missense Mutation |
ATG,GTG |
M265V |
NP_001624.1 |
- Gene
- ZNF618
- Gene Name
- zinc finger protein 618
There are no transcripts associated with this gene.
View Full Product Details