Product Details

SNP ID
rs147724058
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:3825513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTTTGTGTTTTGTTTCTGTTTT[G/T]AAGGGGAATGACAATCTCTGTTTGG
Phenotype
MIM: 610192
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GLIS3 PubMed Links

Gene Details

Gene
GLIS3
Gene Name
GLIS family zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042413.1 5616 UTR 3 NP_001035878.1
NM_152629.3 5616 UTR 3 NP_689842.3
XM_005251386.4 5616 UTR 3 XP_005251443.1
XM_005251387.4 5616 UTR 3 XP_005251444.1
XM_005251388.4 5616 UTR 3 XP_005251445.1
XM_005251389.4 5616 Intron XP_005251446.1
XM_006716731.3 5616 Intron XP_006716794.1
XM_011517763.2 5616 UTR 3 XP_011516065.1
XM_011517764.2 5616 UTR 3 XP_011516066.1
XM_011517765.2 5616 Intron XP_011516067.1
XM_011517766.2 5616 UTR 3 XP_011516068.1
XM_011517767.2 5616 UTR 3 XP_011516069.1
XM_011517769.2 5616 Intron XP_011516071.1
XM_017014361.1 5616 UTR 3 XP_016869850.1

View Full Product Details