Product Details

SNP ID
rs147812056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:79581821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCCTGAACTTTAGGCTCATAC[A/G]CAGAACCTCTTGAGTTGGGGGATTC
Phenotype
MIM: 605132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLE4 PubMed Links

Gene Details

Gene
TLE4
Gene Name
transducin like enhancer of split 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282748.1 Intron NP_001269677.1
NM_001282749.1 Intron NP_001269678.1
NM_001282753.1 Intron NP_001269682.1
NM_001282760.1 Intron NP_001269689.1
NM_007005.4 Intron NP_008936.2
XM_005252177.1 Intron XP_005252234.1
XM_006717264.1 Intron XP_006717327.1
XM_006717268.1 Intron XP_006717331.1
XM_011518952.1 Intron XP_011517254.1
XM_011518953.1 Intron XP_011517255.1
XM_011518954.1 Intron XP_011517256.1
XM_011518955.1 Intron XP_011517257.1
XM_011518956.1 Intron XP_011517258.1
XM_011518957.1 Intron XP_011517259.1
XM_011518958.1 Intron XP_011517260.1
XM_011518959.1 Intron XP_011517261.1
XM_011518960.1 Intron XP_011517262.1
XM_011518961.1 Intron XP_011517263.1
XM_011518962.1 Intron XP_011517264.1
XM_011518963.1 Intron XP_011517265.1
XM_011518964.1 Intron XP_011517266.1
XM_011518965.1 Intron XP_011517267.1
XM_011518966.1 Intron XP_011517268.1
XM_011518967.1 Intron XP_011517269.1
XM_011518968.1 Intron XP_011517270.1
XM_011518969.1 Intron XP_011517271.1
XM_011518970.2 Intron XP_011517272.1
XM_011518972.2 Intron XP_011517274.1
XM_017015067.1 Intron XP_016870556.1
XM_017015068.1 Intron XP_016870557.1
XM_017015069.1 Intron XP_016870558.1
XM_017015070.1 Intron XP_016870559.1
XM_017015071.1 Intron XP_016870560.1
XM_017015072.1 Intron XP_016870561.1
XM_017015073.1 Intron XP_016870562.1
XM_017015074.1 Intron XP_016870563.1
XM_017015075.1 Intron XP_016870564.1
XM_017015076.1 Intron XP_016870565.1
XM_017015077.1 Intron XP_016870566.1
XM_017015078.1 Intron XP_016870567.1
XM_017015079.1 Intron XP_016870568.1
XM_017015080.1 Intron XP_016870569.1
XM_017015081.1 Intron XP_016870570.1
XM_017015082.1 Intron XP_016870571.1
XM_017015083.1 Intron XP_016870572.1
XM_017015084.1 Intron XP_016870573.1
XM_017015085.1 Intron XP_016870574.1
XM_017015086.1 Intron XP_016870575.1

View Full Product Details