Product Details

SNP ID
rs147868113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127925161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCCCCTCTCATCCTCGTGGAGA[C/T]GTTGGAAGGCTATCAGGAGGCTGTA
Phenotype
MIM: 603564 MIM: 612865 MIM: 606378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPM2 PubMed Links

Gene Details

Gene
DPM2
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
There are no transcripts associated with this gene.

Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135219.1 1956 Missense Mutation CAT,CGT H288R NP_001128691.1
NM_173492.1 1956 Missense Mutation CAT,CGT H85R NP_775763.1
XM_017014279.1 1956 Silent Mutation ACA,ACG T333T XP_016869768.1
Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
There are no transcripts associated with this gene.

View Full Product Details