Product Details
- SNP ID
-
rs148215906
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:130668168 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCATCTGTGCCCAGGTGTTCAATG[A/G]GGATGGCACGGTGCGCTACTTCATC
- Phenotype
-
MIM: 616458
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM12
PubMed Links
Gene Details
- Gene
- PRDM12
- Gene Name
- PR domain 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021619.2 |
485 |
Missense Mutation |
GAG,GGG |
E142G |
NP_067632.2 |
View Full Product Details