Product Details

SNP ID
rs148409496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121567171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATAAATCAGGTGGGGCCAGGG[A/G]CTGGGGGCCCACACGCCATGGAGCC
Phenotype
MIM: 609205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAB2IP PubMed Links

Gene Details

Gene
DAB2IP
Gene Name
DAB2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032552.3 289 UTR 5 NP_115941.2
NM_138709.2 289 Intron NP_619723.1
XM_005251719.4 289 Intron XP_005251776.1
XM_005251721.1 289 UTR 5 XP_005251778.1
XM_005251724.4 289 Intron XP_005251781.1
XM_011518264.2 289 Intron XP_011516566.1
XM_011518265.2 289 Intron XP_011516567.1
XM_011518266.2 289 Intron XP_011516568.1
XM_011518267.2 289 Intron XP_011516569.1
XM_011518270.2 289 Intron XP_011516572.1
XM_011518271.2 289 Intron XP_011516573.1
XM_017014298.1 289 Intron XP_016869787.1
XM_017014299.1 289 Intron XP_016869788.1
XM_017014300.1 289 Intron XP_016869789.1

View Full Product Details