Product Details

SNP ID
rs148502447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83661503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAAGCAAATAAACTGGACAGA[G/T]GCAGGACAAAATCTGGTCACCCAAC
Phenotype
MIM: 605046
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBQLN1 PubMed Links

Gene Details

Gene
UBQLN1
Gene Name
ubiquilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013438.4 2544 UTR 3 NP_038466.2
NM_053067.2 2544 UTR 3 NP_444295.1
XM_005251948.2 2544 Intron XP_005252005.1

View Full Product Details