Product Details
- SNP ID
-
rs148578435
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35700198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTCAAGGATTTCTACCTTGCAA[A/G]CCCGAAGCATATCTGCAATAGCACG
- Phenotype
-
MIM: 186745
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLN1
PubMed Links
Gene Details
- Gene
- TLN1
- Gene Name
- talin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006289.3 |
7007 |
Missense Mutation |
GCT,GTT |
A2218V |
NP_006280.3 |
View Full Product Details