Product Details

SNP ID
rs149627939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122553540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGCTATATTTCCTCCTTATGT[C/T]TGGTGGCCTTGACAACTGCCTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1L8 PubMed Links

Gene Details

Gene
OR1L8
Gene Name
olfactory receptor family 1 subfamily L member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004454.1 371 Intron NP_001004454.1
XM_017014284.1 371 Intron XP_016869773.1
XM_017014285.1 371 Intron XP_016869774.1
XM_017014286.1 371 Intron XP_016869775.1
Gene
OR1N2
Gene Name
olfactory receptor family 1 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004457.1 371 Missense Mutation TCT,TTT S124F NP_001004457.1

View Full Product Details